The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033306.1 | NP_001028478.1 | 4 - 5 | 1778 | 69 |
GenBank mRNA | AK145414.1 | - | 4 - 5 | 637 | 69 |
BC028832.1 | - | 3 - 4 | 640 | 69 | |
BC048850.1 | - | 3 - 4 | 645 | 69 | |
BC058986.1 | - | 4 - 5 | 1280 | 69 |