The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017476.2 | NP_059504.2 | 3 - 4 | 198 | 84 |
GenBank mRNA | AB028921.1 | - | 3 - 4 | 191 | 84 |
AK011325.1 | - | 3 - 4 | 203 | 84 | |
AK166526.1 | - | 3 - 4 | 216 | 84 | |
AK206808.1 | - | 2 - 3 | 71 | 84 | |
BC010401.1 | - | 3 - 4 | 248 | 84 | |
BC046801.1 | - | 3 - 4 | 187 | 84 | |
BC051015.1 | - | 3 - 4 | 198 | 84 |