Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pramef8
Entrez Gene ID
242736
Gene Name
PRAME family member 8
Gene Aliases
4732496O08Rik, AU023234
Chromosome Location
Chr.4: 143412309 - 143421091 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC056931.1 - 1 - 2 103 70

View Full Product Details