The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153097.2 | NP_694737.1 | 2 - 2 | 2669 | 112 |
GenBank mRNA | AK135841.1 | - | 2 - 2 | 2567 | 112 |