The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290684.1 | NP_001277613.1 | 9 - 10 | 4214 | 100 |
NM_001290685.1 | NP_001277614.1 | 5 - 6 | 1539 | 100 | |
NM_001290686.1 | NP_001277615.1 | 5 - 6 | 1346 | 100 | |
NM_001290687.1 | NP_001277616.1 | 5 - 6 | 1528 | 100 | |
NM_172441.3 | NP_766029.2 | 9 - 10 | 4232 | 100 | |
GenBank mRNA | AK029338.1 | - | 5 - 6 | 1220 | 100 |
AK032256.1 | - | 5 - 6 | 1221 | 100 | |
AK049833.1 | - | 1 - 2 | 200 | 100 | |
AK049850.1 | - | 5 - 6 | 1278 | 100 | |
BC092373.1 | - | 5 - 6 | 1534 | 100 | |
DQ358971.1 | - | 9 - 10 | 4192 | 100 | |
EF071946.1 | - | 9 - 10 | 4232 | 100 |