Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vill
Entrez Gene ID
22351
Gene Name
villin-like
Gene Aliases
Villp
Chromosome Location
Chr.9: 119052729 - 119071525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164567.1 NP_001158039.1 14 - 15 1717 92
NM_011700.2 NP_035830.2 14 - 15 1717 92
GenBank mRNA AJ344341.1 - 14 - 15 1705 92
AK028229.1 - 14 - 15 1718 92
AK166823.1 - 13 - 14 1949 92
BC022664.1 - 5 - 6 646 92

View Full Product Details