Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vill
Entrez Gene ID
22351
Gene Name
villin-like
Gene Aliases
Villp
Chromosome Location
Chr.9: 119052729 - 119071525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164567.1 NP_001158039.1 1 - 2 54 53
NM_011700.2 NP_035830.2 1 - 2 54 53
GenBank mRNA AK028229.1 - 1 - 2 55 53
AK166823.1 - 1 - 2 55 53
BB611096.1 - 1 - 2 55 53
CN670511.1 - 1 - 2 30 53

View Full Product Details