The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019701.2 | NP_062675.2 | 9 - 10 | 921 | 97 |
GenBank mRNA | AF124847.1 | - | 9 - 10 | 914 | 97 |
AK146876.1 | - | 8 - 9 | 984 | 97 | |
BC019983.1 | - | 9 - 10 | 947 | 97 |