Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pink1
Entrez Gene ID
68943
Gene Name
PTEN induced putative kinase 1
Gene Aliases
1190006F07Rik, AU042772, AW557854, BRPK, mFLJ00387
Chromosome Location
Chr.4: 138313409 - 138326296 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC018594.1 - 5 - 6 1159 70
BC054349.1 - 1 - 2 174 70

View Full Product Details