Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctrc
Entrez Gene ID
76701
Gene Name
chymotrypsin C (caldecrin)
Gene Aliases
1810044E12Rik
Chromosome Location
Chr.4: 141838239 - 141846359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033875.2 NP_001029047.1 7 - 8 880 62
GenBank mRNA AK007772.1 - 6 - 7 708 62
BC103715.1 - 7 - 8 878 62

View Full Product Details