Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fhad1
Entrez Gene ID
329977
Gene Name
forkhead-associated (FHA) phosphopeptide binding domain 1
Gene Aliases
2900090M10Rik, B230311F11
Chromosome Location
Chr.4: 141890430 - 142015124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177868.4 NP_808536.2 19 - 20 2603 59
GenBank mRNA AK148490.1 - 4 - 5 688 59
BC038692.1 - 1 - 2 186 59

View Full Product Details