Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab3a
Entrez Gene ID
19339
Gene Name
RAB3A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.8: 70754666 - 70758686 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009001.6 NP_033027.1 1 - 2 189 183
GenBank mRNA AK161975.1 - 1 - 2 158 183
BY097011.1 - 1 - 2 190 183

View Full Product Details