Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl26
Entrez Gene ID
234378
Gene Name
kelch-like 26
Gene Aliases
C630013N10Rik, Klkl26
Chromosome Location
Chr.8: 70450221 - 70476988 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC023909.1 - 1 - 2 106 86

View Full Product Details