Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Arl13b
- Entrez Gene ID
-
68146
- Gene Name
- ADP-ribosylation factor-like 13B
- Gene Aliases
-
A530097K21Rik, A930014M17Rik, Arl2l1, C530009C10Rik, hnn
- Chromosome Location
-
Chr.16: 62793678 - 62847055 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_026577.3 |
NP_080853.3 |
7 - 8 |
1315 |
106 |
View Full Product Details