The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015759.2 | NP_056574.2 | 15 - 16 | 1903 | 77 |
GenBank mRNA | AF017369.1 | - | 15 - 16 | 1938 | 77 |
AK157835.1 | - | - | 1903 | 77 | |
AK165413.1 | - | 15 - 16 | 2313 | 77 | |
BC132340.1 | - | 13 - 14 | 1722 | 77 | |
BC137954.1 | - | 13 - 14 | 1723 | 77 |