Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hist3h2ba
Entrez Gene ID
78303
Gene Name
histone cluster 3, H2ba
Gene Aliases
1500011O09Rik, AI413321
Chromosome Location
Chr.11: 58948911 - 58949372 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030082.4 NP_084358.1 1 - 1 48 80
GenBank mRNA AK018765.1 - 1 - 1 49 80
BC051921.1 - 1 - 1 39 80

View Full Product Details