Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Iba57
Entrez Gene ID
216792
Gene Name
IBA57, iron-sulfur cluster assembly homolog (S. cerevisiae)
Gene Aliases
4930543L23Rik, A230051G13Rik, C1orf69
Chromosome Location
Chr.11: 59155368 - 59164591 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK172525.1 - 2 - 3 696 163
BC094909.1 - 2 - 3 735 163

View Full Product Details