Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cplx2
Entrez Gene ID
12890
Gene Name
complexin 2
Gene Aliases
921-L, AI413745, AW492120
Chromosome Location
Chr.13: 54371349 - 54383917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009946.3 NP_034076.1 3 - 4 526 64
GenBank mRNA CX241609.1 - 3 - 4 526 64
D38613.1 - 2 - 3 295 64

View Full Product Details