Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxw17
Entrez Gene ID
109082
Gene Name
F-box and WD-40 domain protein 17
Gene Aliases
1110064L07Rik, C85886
Chromosome Location
Chr.13: 50417834 - 50433781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175401.3 NP_780610.2 8 - 9 1192 59
GenBank mRNA AK155157.1 - 9 - 10 1706 59
BC040428.1 - 8 - 9 1192 59

View Full Product Details