Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spock1
Entrez Gene ID
20745
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan 1
Gene Aliases
Spock, Ticn1, testican
Chromosome Location
Chr.13: 57421195 - 57908332 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166463.1 NP_001159935.1 10 - 11 1268 63
NM_001166466.1 NP_001159938.1 11 - 12 1274 63
NM_009262.3 NP_033288.2 11 - 12 1277 63
GenBank mRNA AK051191.1 - 10 - 11 1228 63
BC075625.1 - 11 - 12 1220 63
BC158020.1 - 11 - 12 1181 63
X92864.1 - 11 - 12 1269 63

View Full Product Details