Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Car9
Entrez Gene ID
230099
Gene Name
carbonic anhydrase 9
Gene Aliases
CAIX, Ca9, MN/CA9
Chromosome Location
Chr.4: 43507026 - 43513810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139305.2 NP_647466.2 7 - 8 1033 79
GenBank mRNA AJ245857.1 - 7 - 8 1035 79
AK136579.1 - 7 - 8 1034 79
AK137719.1 - 7 - 8 1035 79
BC120544.1 - 7 - 8 1026 79
BC120546.1 - 7 - 8 1026 79

View Full Product Details