Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Car9
Entrez Gene ID
230099
Gene Name
carbonic anhydrase 9
Gene Aliases
CAIX, Ca9, MN/CA9
Chromosome Location
Chr.4: 43507026 - 43513810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EF122497.1 - 6 - 7 875 64

View Full Product Details