Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd32
Entrez Gene ID
105377
Gene Name
ankyrin repeat domain 32
Gene Aliases
2700017A04Rik, AW545819, Brctd1, Brctx, C730024G01Rik
Chromosome Location
Chr.13: 77043088 - 77135468 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134071.3 NP_598832.3 1 - 2 118 103
GenBank mRNA AK012253.1 - 1 - 2 111 103
AK089501.1 - 1 - 2 118 103
AK171473.1 - 1 - 2 118 103
BC061188.1 - 1 - 2 99 103
BC100362.1 - 1 - 2 90 103

View Full Product Details