Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Golgb1
Entrez Gene ID
224139
Gene Name
golgi autoantigen, golgin subfamily b, macrogolgin 1
Gene Aliases
4930428L02Rik, 628101, 6330407A06Rik, AU042952, C130074L01Rik, F730017E11Rik, Gm6840, mKIAA4151
Chromosome Location
Chr.16: 36875173 - 36933085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030035.1 NP_084311.1 21 - 22 9765 65
GenBank mRNA AK081762.1 - 2 - 3 168 65
AK220188.1 - 9 - 10 5620 65
BC040461.1 - 9 - 10 3085 65
BC060254.1 - 8 - 9 2189 65
BC145221.1 - 19 - 20 2707 65
BC150731.1 - 21 - 22 9742 65

View Full Product Details