Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tctex1d2
- Entrez Gene ID
-
66061
- Gene Name
- Tctex1 domain containing 2
- Gene Aliases
-
0610012D17Rik, 2410011K15Rik
- Chromosome Location
-
Chr.16: 32419686 - 32429099 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC053332.1 |
- |
1 - 2 |
128 |
78 |
View Full Product Details