Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Faf2
- Entrez Gene ID
-
76577
- Gene Name
- Fas associated factor family member 2
- Gene Aliases
-
2210404D11Rik, AI462440, Ubxd8, mKIAA0887
- Chromosome Location
-
Chr.13: 54621775 - 54664063 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK151617.1 |
- |
4 - 5 |
310 |
116 |
View Full Product Details