Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Kazn
- Entrez Gene ID
-
71529
- Gene Name
- kazrin, periplakin interacting protein
- Gene Aliases
-
2310007B04Rik, 9030409G11Rik, AI256607, AI666764, AW061099, Kaz, Kazrin
- Chromosome Location
-
Chr.4: 142102390 - 142239401 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK173090.1 |
- |
11 - 12 |
2046 |
81 |
View Full Product Details