Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a34
Entrez Gene ID
384071
Gene Name
solute carrier family 25, member 34
Gene Aliases
Gm1369
Chromosome Location
Chr.4: 141618824 - 141623993 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013780.1 NP_001013802.1 3 - 4 776 112
GenBank mRNA AK142924.1 - 3 - 4 777 113

View Full Product Details