Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nktr
Entrez Gene ID
18087
Gene Name
natural killer tumor recognition sequence
Gene Aliases
5330401F18Rik, D9Wsu172e
Chromosome Location
Chr.9: 121718181 - 121756842 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010918.2 NP_035048.3 12 - 13 1305 66
GenBank mRNA AK035582.1 - 11 - 12 1131 66
AK144214.1 - 12 - 13 1216 66
BC048414.1 - 13 - 14 1295 66
L04289.2 - 12 - 13 1310 66

View Full Product Details