Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Igf2bp2
Entrez Gene ID
319765
Gene Name
insulin-like growth factor 2 mRNA binding protein 2
Gene Aliases
C330012H03Rik, IMP-2, Imp2, Neilsen
Chromosome Location
Chr.16: 22059009 - 22163299 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183029.2 NP_898850.2 7 - 8 1133 109
GenBank mRNA AK044984.1 - 6 - 7 649 109
AK049196.1 - 7 - 8 830 109
AK077404.1 - 7 - 8 813 109
AK159654.1 - 7 - 8 866 109
AK170531.1 - 6 - 7 637 109
AY531659.1 - 7 - 8 797 109
BC023758.1 - 7 - 8 831 109
BC054552.1 - 7 - 8 853 109

View Full Product Details