The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030263.5 | NP_084539.2 | 6 - 7 | 690 | 106 |
NM_177698.4 | NP_808366.2 | 9 - 10 | 2454 | 106 | |
GenBank mRNA | AB220685.1 | - | 9 - 10 | 2454 | 106 |
AK028684.1 | - | 9 - 10 | 2263 | 106 | |
AK161498.1 | - | 6 - 7 | 676 | 106 | |
BC052365.1 | - | 6 - 7 | 654 | 106 |