Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amtn
Entrez Gene ID
71421
Gene Name
amelotin
Gene Aliases
5430427O21Rik
Chromosome Location
Chr.5: 88376108 - 88385916 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027793.1 NP_082069.1 2 - 3 140 104
GenBank mRNA AK017352.1 - 2 - 3 140 104

View Full Product Details