Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr48
Entrez Gene ID
67561
Gene Name
WD repeat domain 48
Gene Aliases
8430408H12Rik, Uaf1, mKIAA1449
Chromosome Location
Chr.9: 119894895 - 119926586 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026236.3 NP_080512.1 12 - 13 1301 133
GenBank mRNA AK018396.1 - 12 - 13 1304 133
AK030041.1 - 12 - 13 1302 133
AK031244.1 - 12 - 13 1314 133
AK033631.1 - 12 - 13 1300 133
AK035273.1 - 12 - 13 1303 133
AK035864.1 - 12 - 13 1303 133
AK043970.1 - 12 - 13 1303 133
AK122511.1 - 11 - 12 2443 133
AK128953.1 - 12 - 13 1301 133
BC006679.1 - 11 - 12 1164 133
BC021348.1 - 2 - 3 1491 133
BC040795.1 - 8 - 9 832 133
BC050106.1 - 12 - 13 1319 134
BC062967.1 - 12 - 13 1305 133

View Full Product Details