Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr48
Entrez Gene ID
67561
Gene Name
WD repeat domain 48
Gene Aliases
8430408H12Rik, Uaf1, mKIAA1449
Chromosome Location
Chr.9: 119894895 - 119926586 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026236.3 NP_080512.1 17 - 18 1761 70
GenBank mRNA AK030041.1 - 17 - 18 1762 70
AK031244.1 - 17 - 18 1776 70
AK033631.1 - 17 - 18 1760 70
AK035273.1 - 17 - 18 1763 70
AK043970.1 - 17 - 18 1763 70
AK122511.1 - 16 - 17 2903 70
AK128953.1 - 17 - 18 1761 70
BC006679.1 - 16 - 17 1624 70
BC040795.1 - 13 - 14 1292 70
BC048155.1 - 17 - 18 1708 70
BC050106.1 - 17 - 18 1779 70
BC062967.1 - 17 - 18 1765 70

View Full Product Details