The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033209.3 | NP_001028381.1 | 15 - 16 | 1562 | 56 |
NM_001199568.1 | NP_001186497.1 | 16 - 17 | 1610 | 56 | |
GenBank mRNA | AK143476.1 | - | 15 - 16 | 1562 | 56 |
AK164186.1 | - | 16 - 17 | 1404 | 56 | |
AK165440.1 | - | 15 - 16 | 1359 | 56 | |
AK165518.1 | - | 15 - 16 | 1359 | 56 | |
AK165759.1 | - | 16 - 17 | 1406 | 56 | |
AK167735.1 | - | 6 - 7 | 582 | 56 | |
AK170197.1 | - | 14 - 15 | 1307 | 56 | |
BC025442.1 | - | 15 - 16 | 1312 | 56 | |
BC026406.1 | - | 3 - 4 | 274 | 56 | |
BC080743.1 | - | 15 - 16 | 1336 | 56 | |
BC138244.1 | - | 15 - 16 | 1511 | 56 | |
BC138247.1 | - | 15 - 16 | 1511 | 56 |