Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il12rb1
Entrez Gene ID
16161
Gene Name
interleukin 12 receptor, beta 1
Gene Aliases
CD212, IL-12R[b]
Chromosome Location
Chr.8: 70808360 - 70821424 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008353.2 NP_032379.2 5 - 6 677 67
GenBank mRNA AK137577.1 - 5 - 6 678 67
AK171937.1 - 5 - 6 654 67

View Full Product Details