The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048146.2 | NP_001041611.1 | 3 - 4 | 776 | 144 |
NM_001286508.1 | NP_001273437.1 | 3 - 4 | 694 | 144 | |
NM_013727.4 | NP_038755.1 | 3 - 4 | 776 | 144 | |
GenBank mRNA | AB007141.1 | - | 3 - 4 | 612 | 144 |
AK015942.1 | - | 3 - 4 | 597 | 144 | |
AK030076.1 | - | 3 - 4 | 608 | 144 | |
AK134714.1 | - | 3 - 4 | 679 | 144 | |
AK153477.1 | - | 3 - 4 | 740 | 144 | |
AK165549.1 | - | 3 - 4 | 662 | 144 | |
AK167783.1 | - | 3 - 4 | 720 | 144 | |
BC006851.1 | - | 3 - 4 | 671 | 144 |