Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mau2
Entrez Gene ID
74549
Gene Name
MAU2 chromatid cohesion factor homolog (C. elegans)
Gene Aliases
9130404D08Rik, A930019L04Rik, C77863, C79014, Mau-2, mKIAA0892
Chromosome Location
Chr.8: 70014527 - 70042734 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167939.1 NP_001161411.1 15 - 16 1503 69
NM_028993.4 NP_083269.4 15 - 16 1500 69
GenBank mRNA AK018662.1 - 15 - 16 1468 69
AK034837.1 - 15 - 16 1469 69
AK039196.1 - 15 - 16 1469 69
AK049277.1 - 15 - 16 1461 69
AK081967.1 - 15 - 16 1501 69
AK162318.1 - 13 - 14 2845 69
BC023401.1 - 5 - 6 354 69
BC125568.1 - 15 - 16 1500 69
BC132139.1 - 15 - 16 1500 69
BC144990.1 - 15 - 16 1503 69

View Full Product Details