The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023281.1 | NP_075770.1 | 10 - 11 | 1466 | 74 |
GenBank mRNA | AF095938.1 | - | 8 - 9 | 1215 | 74 |
AK010561.1 | - | 10 - 11 | 1455 | 74 | |
AK029520.1 | - | 10 - 11 | 1466 | 74 | |
AK034928.1 | - | 10 - 11 | 1467 | 74 | |
AK049590.1 | - | 10 - 11 | 1453 | 74 | |
AK050475.1 | - | 10 - 11 | 1453 | 74 | |
AK075990.1 | - | 10 - 11 | 1450 | 74 | |
AK145923.1 | - | 10 - 11 | 1464 | 74 | |
AK147286.1 | - | 10 - 11 | 1453 | 74 | |
AK147624.1 | - | 10 - 11 | 1453 | 74 | |
AK153085.1 | - | 10 - 11 | 1473 | 74 | |
AK162148.1 | - | 10 - 11 | 1449 | 74 | |
AK169254.1 | - | 10 - 11 | 1449 | 74 | |
BC011301.1 | - | 10 - 11 | 1426 | 74 | |
BC031849.1 | - | 10 - 11 | 1474 | 74 | |
DQ402975.1 | - | 8 - 9 | 1260 | 74 |