Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sdha
Entrez Gene ID
66945
Gene Name
succinate dehydrogenase complex, subunit A, flavoprotein (Fp)
Gene Aliases
1500032O14Rik, 2310034D06Rik, 4921513A11, C81073, FP, SDH2, SDHF
Chromosome Location
Chr.13: 74322255 - 74350240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023281.1 NP_075770.1 4 - 5 489 78
GenBank mRNA AF095938.1 - 2 - 3 238 78
AK010561.1 - 4 - 5 478 78
AK029520.1 - 4 - 5 489 78
AK034928.1 - 4 - 5 490 78
AK049590.1 - 4 - 5 476 78
AK050475.1 - 4 - 5 476 78
AK075990.1 - 4 - 5 473 78
AK145923.1 - 4 - 5 487 78
AK147286.1 - 4 - 5 476 78
AK147624.1 - 4 - 5 476 78
AK153085.1 - 4 - 5 496 78
AK162148.1 - 4 - 5 472 78
AK169254.1 - 4 - 5 472 78
AK209906.1 - 2 - 3 297 78
AK215982.1 - 2 - 3 303 78
AK216171.1 - 2 - 3 291 78
BC011301.1 - 4 - 5 449 78
BC031849.1 - 4 - 5 497 78
DQ402975.1 - 2 - 3 283 78

View Full Product Details