Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trip13
Entrez Gene ID
69716
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
2410002G23Rik, D13Ertd328e
Chromosome Location
Chr.13: 73912462 - 73937767 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027182.2 NP_081458.1 4 - 5 625 71
GenBank mRNA AK142463.1 - - 598 71
AK146877.1 - 4 - 5 626 71
AK151568.1 - 4 - 5 561 71
BC064315.1 - 4 - 5 548 71
BC126946.1 - 4 - 5 524 71

View Full Product Details