Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trip13
Entrez Gene ID
69716
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
2410002G23Rik, D13Ertd328e
Chromosome Location
Chr.13: 73912462 - 73937767 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027182.2 NP_081458.1 9 - 10 1051 86
GenBank mRNA AK010336.1 - 9 - 10 1013 86
AK142463.1 - - 1024 86
AK146877.1 - 9 - 10 1052 86
AK151568.1 - 9 - 10 987 86
BC023834.1 - 3 - 4 259 86
BC064315.1 - 9 - 10 974 86
BC126946.1 - 9 - 10 950 86

View Full Product Details