Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sfxn1
Entrez Gene ID
14057
Gene Name
sideroflexin 1
Gene Aliases
2810002O05Rik, A930015P12Rik, f
Chromosome Location
Chr.13: 54071824 - 54108346 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027324.5 NP_081600.1 4 - 5 532 90
GenBank mRNA AF325260.1 - 4 - 5 510 90
AK012650.1 - 4 - 5 531 90
AK151110.1 - 4 - 5 478 90
AK179717.1 - 3 - 4 317 90
BC005743.1 - 4 - 5 489 90

View Full Product Details