Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cetn3
Entrez Gene ID
12626
Gene Name
centrin 3
Gene Aliases
MmCEN3
Chromosome Location
Chr.13: 81783247 - 81797157 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007684.3 NP_031710.1 1 - 2 61 144
GenBank mRNA AK003473.1 - 1 - 2 78 143
AK004532.1 - 1 - 2 73 144
AK006289.1 - 1 - 2 76 144
AK006891.1 - 1 - 2 61 144
AK011821.1 - 1 - 2 54 144
AK017505.1 - 1 - 2 101 144
BC002162.1 - 1 - 2 95 144
BC054097.1 - 1 - 2 97 144

View Full Product Details