Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cetn3
Entrez Gene ID
12626
Gene Name
centrin 3
Gene Aliases
MmCEN3
Chromosome Location
Chr.13: 81783247 - 81797157 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007684.3 NP_031710.1 4 - 5 506 136
GenBank mRNA AK003473.1 - 4 - 5 522 136
AK004532.1 - 4 - 5 518 136
AK006289.1 - 4 - 5 521 136
AK006891.1 - 4 - 5 506 136
AK017505.1 - 4 - 5 546 136
BC002162.1 - 4 - 5 540 136
BC054097.1 - 4 - 5 542 136

View Full Product Details