Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf2
Entrez Gene ID
18676
Gene Name
PHD finger protein 2
Gene Aliases
GRC5
Chromosome Location
Chr.13: 48801750 - 48870885 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011078.3 NP_035208.2 3 - 4 423 68
GenBank mRNA AB221591.1 - 3 - 4 307 68
AF043726.1 - 3 - 4 423 68
AK172995.1 - 1 - 2 130 68
BC051633.1 - 3 - 4 421 68

View Full Product Details