The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033429.2 | NP_001028601.1 | 2 - 2 | 1282 | 124 |
GenBank mRNA | AK136018.1 | - | 2 - 2 | 1284 | 124 |
AK162165.1 | - | 1 - 1 | 125 | 124 | |
AK163251.1 | - | 1 - 1 | 639 | 124 | |
BC139219.1 | - | 1 - 1 | 989 | 124 | |
BC139221.1 | - | 1 - 1 | 989 | 124 |