The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009254.3 | NP_033280.1 | 1 - 2 | 162 | 72 |
GenBank mRNA | AK033731.1 | - | 1 - 2 | 58 | 72 |
AK167295.1 | - | 1 - 2 | 57 | 72 | |
AK169145.1 | - | 1 - 2 | 55 | 72 | |
BC057950.1 | - | 1 - 2 | 42 | 72 | |
U25844.1 | - | 1 - 2 | 57 | 72 |