The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183319.2 | NP_899142.2 | 1 - 2 | 360 | 76 |
GenBank mRNA | AY534249.1 | - | 1 - 2 | 360 | 76 |
BC150896.1 | - | 1 - 2 | 396 | 76 | |
BC150897.1 | - | 1 - 2 | 396 | 76 |