The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177937.1 | NP_001171408.1 | 1 - 2 | 571 | 118 |
NM_001177938.1 | NP_001171409.1 | 1 - 2 | 571 | 118 | |
NM_001177939.1 | NP_001171410.1 | 1 - 2 | 571 | 118 | |
NM_001177940.1 | NP_001171411.1 | 1 - 2 | 571 | 118 | |
NM_001177941.1 | NP_001171412.1 | 1 - 2 | 571 | 118 | |
NM_001177942.1 | NP_001171413.1 | 1 - 2 | 571 | 118 | |
NM_001177943.1 | NP_001171414.1 | 1 - 2 | 571 | 118 | |
NM_001177944.1 | NP_001171415.1 | 1 - 2 | 571 | 118 | |
NM_010099.2 | NP_034229.1 | 1 - 2 | 571 | 118 | |
GenBank mRNA | AF004435.1 | - | 1 - 2 | 537 | 118 |
AF016627.1 | - | 1 - 2 | 571 | 118 | |
AF016630.1 | - | 1 - 2 | 571 | 118 | |
AJ243657.1 | - | 1 - 2 | 558 | 118 | |
AJ243658.1 | - | 1 - 2 | 519 | 118 | |
AK137551.1 | - | 1 - 2 | 480 | 118 | |
BC125387.1 | - | 1 - 2 | 462 | 118 | |
BC144790.1 | - | 1 - 2 | 462 | 118 | |
BC144791.1 | - | 1 - 2 | 462 | 118 | |
DQ168876.1 | - | 1 - 2 | 571 | 118 | |
DQ168877.1 | - | 1 - 2 | 571 | 118 | |
DQ168878.1 | - | 1 - 2 | 571 | 118 | |
DQ168879.1 | - | 1 - 2 | 571 | 118 | |
DQ168880.1 | - | 1 - 2 | 571 | 118 | |
DQ168881.1 | - | 1 - 2 | 571 | 118 |